Learn More
1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6384503
Description
1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
1-[3-(Dimethylamino)propyl]-3-ethylurea | |
C8H19N3O | |
MFCD00027752 | |
4961600 | |
Soluble in water. | |
CCNC(=O)NCCCN(C)C | |
173.26 | |
173.26 |
32897-26-0 | |
CH3CH2NHCONH(CH2)3N(CH3)2 | |
1 g | |
1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine | |
NGJUYARYEXGDNN-UHFFFAOYSA-N | |
1-[3-(dimethylamino)propyl]-3-ethylurea | |
4196313 | |
97% |
Chemical Identifiers
32897-26-0 | |
173.26 | |
NGJUYARYEXGDNN-UHFFFAOYSA-N | |
4196313 | |
CCNC(=O)NCCCN(C)C |
C8H19N3O | |
MFCD00027752 | |
1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine | |
1-[3-(dimethylamino)propyl]-3-ethylurea |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only