Learn More
1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%, Thermo Scientific Chemicals
$102.04 - $1078.86
Chemical Identifiers
CAS | 32897-26-0 |
---|---|
Molecular Formula | C8H19N3O |
Molecular Weight (g/mol) | 173.26 |
MDL Number | MFCD00027752 |
InChI Key | NGJUYARYEXGDNN-UHFFFAOYSA-N |
Synonym | 1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine |
PubChem CID | 4196313 |
IUPAC Name | 1-[3-(dimethylamino)propyl]-3-ethylurea |
SMILES | CCNC(=O)NCCCN(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63845MD
|
Thermo Scientific Chemicals
H63845MD |
250 mg |
Each for $102.04
|
|
|||||
AAH6384503
|
Thermo Scientific Chemicals
H6384503 |
1 g |
Each for $281.23
|
|
|||||
AAH6384506
|
Thermo Scientific Chemicals
H6384506 |
5 g |
Each for $1,078.86
|
|
|||||
Description
1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
32897-26-0 | |
173.26 | |
NGJUYARYEXGDNN-UHFFFAOYSA-N | |
4196313 | |
CCNC(=O)NCCCN(C)C |
C8H19N3O | |
MFCD00027752 | |
1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine | |
1-[3-(dimethylamino)propyl]-3-ethylurea |
Specifications
32897-26-0 | |
CH3CH2NHCONH(CH2)3N(CH3)2 | |
250 mg | |
1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine | |
NGJUYARYEXGDNN-UHFFFAOYSA-N | |
1-[3-(dimethylamino)propyl]-3-ethylurea | |
4196313 | |
97% |
C8H19N3O | |
MFCD00027752 | |
4961600 | |
Soluble in water. | |
CCNC(=O)NCCCN(C)C | |
173.26 | |
173.26 | |
1-[3-(Dimethylamino)propyl]-3-ethylurea |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only