missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 171440010

 View more versions of this product

Catalog No. AC171440010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Specifications

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride
98.0
111.0°C to 116.0°C
Authentic
Glass bottle
(CH3)2NCH2CH2CH2N=C=NCH2CH3
1 g
15, 3555
Solubility in water: >200g/l. Other solubilities: soluble in ethanol
Cl.CCN=C=NCCCN(C)C
191.70
191.71
Crystalline Powder
25952-53-8
100.0
White
98% min. (Argentometry)
C8H18ClN3
MFCD00012503
12,199
1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl
FPQQSJJWHUJYPU-UHFFFAOYSA-N
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride
2723939
98+%

Chemical Identifiers

25952-53-8
191.70
FPQQSJJWHUJYPU-UHFFFAOYSA-N
2723939
Cl.CCN=C=NCCCN(C)C
C8H18ClN3
MFCD00012503
1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical

GHS Signal Word: Warning

EINECSNumber : 247-361-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only