Learn More
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals
$55.66 - $5734.31
Chemical Identifiers
CAS | 25952-53-8 |
---|---|
Molecular Formula | C8H18ClN3 |
Molecular Weight (g/mol) | 191.70 |
MDL Number | MFCD00012503 |
InChI Key | FPQQSJJWHUJYPU-UHFFFAOYSA-N |
Synonym | 1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl |
PubChem CID | 2723939 |
IUPAC Name | 3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride |
SMILES | Cl.CCN=C=NCCCN(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC171440010
|
Thermo Scientific Chemicals
171440010 |
1 g | Glass bottle |
Each for $55.66
|
|
|||||
AC171440100
|
Thermo Scientific Chemicals
171440100 |
10 g | Glass Bottle |
Each for $240.33
|
|
|||||
AC171440500
|
Thermo Scientific Chemicals
171440500 |
50 g | Glass Bottle |
Each for $1,011.34
|
|
|||||
AC171441000
|
Thermo Scientific Chemicals
171441000 |
100 g | Glass Bottle |
Each for $1,510.17
|
|
|||||
AC171445000
|
Thermo Scientific Chemicals
171445000 |
500 g | Glass bottle |
Each for $5,734.31
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
25952-53-8 | |
191.70 | |
FPQQSJJWHUJYPU-UHFFFAOYSA-N | |
2723939 | |
Cl.CCN=C=NCCCN(C)C |
C8H18ClN3 | |
MFCD00012503 | |
1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl | |
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride |
Specifications
25952-53-8 | |
100.0 | |
White | |
98% min. (Argentometry) | |
C8H18ClN3 | |
MFCD00012503 | |
12,199 | |
1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl | |
FPQQSJJWHUJYPU-UHFFFAOYSA-N | |
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride | |
2723939 | |
98+% | |
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride |
98.0 | |
111.0°C to 116.0°C | |
Authentic | |
Glass bottle | |
(CH3)2NCH2CH2CH2N=C=NCH2CH3 | |
1 g | |
15, 3555 | |
Solubility in water: >200g/l. Other solubilities: soluble in ethanol | |
Cl.CCN=C=NCCCN(C)C | |
191.70 | |
191.71 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical
GHS Signal Word: Warning
EINECSNumber : 247-361-2
RUO – Research Use Only