missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 334120100

 View more versions of this product

Catalog No. AC334120100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Specifications

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide
22572-40-3
100.0
White
97.5% min. (Argentometry)
C9H20IN3
10 g
3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
JHMDMCSGQQLHNA-UHFFFAOYSA-N
3-(ethyliminomethylideneamino)propyl-trimethylazanium
122243
98%
98%
97.5
92.0°C to 97.0°C
Authentic
Glass bottle
MFCD00011833
15, 3555
Solubility in water: 50g/L
CCN=C=NCCC[N+](C)(C)C
297.18
297.18
Crystalline Powder

Chemical Identifiers

22572-40-3
297.18
JHMDMCSGQQLHNA-UHFFFAOYSA-N
122243
CCN=C=NCCC[N+](C)(C)C
C9H20IN3
MFCD00011833
3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
3-(ethyliminomethylideneamino)propyl-trimethylazanium
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respir

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to d

GHS Signal Word: Danger

EINECSNumber : 245-095-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only