Learn More
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%, Thermo Scientific Chemicals
$270.51 - $270.51
Chemical Identifiers
CAS | 22572-40-3 |
---|---|
Molecular Formula | C9H20IN3 |
Molecular Weight (g/mol) | 297.18 |
MDL Number | MFCD00011833 |
InChI Key | JHMDMCSGQQLHNA-UHFFFAOYSA-N |
Synonym | 3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene |
PubChem CID | 122243 |
IUPAC Name | 3-(ethyliminomethylideneamino)propyl-trimethylazanium |
SMILES | CCN=C=NCCC[N+](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC334120100
|
Thermo Scientific Chemicals
334120100 |
10 g | Glass bottle |
Each for $270.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
22572-40-3 | |
297.18 | |
JHMDMCSGQQLHNA-UHFFFAOYSA-N | |
122243 | |
CCN=C=NCCC[N+](C)(C)C |
C9H20IN3 | |
MFCD00011833 | |
3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene | |
3-(ethyliminomethylideneamino)propyl-trimethylazanium |
Specifications
22572-40-3 | |
100.0 | |
White | |
97.5% min. (Argentometry) | |
C9H20IN3 | |
10 g | |
3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene | |
JHMDMCSGQQLHNA-UHFFFAOYSA-N | |
3-(ethyliminomethylideneamino)propyl-trimethylazanium | |
122243 | |
98% | |
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98% |
97.5 | |
92.0°C to 97.0°C | |
Authentic | |
Glass bottle | |
MFCD00011833 | |
15, 3555 | |
Solubility in water: 50g/L | |
CCN=C=NCCC[N+](C)(C)C | |
297.18 | |
297.18 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respir
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to d
GHS Signal Word: Danger
EINECSNumber : 245-095-1
TSCA : TSCA
RUO – Research Use Only