missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Diphenyl-1,3-propanedione, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1345036

 View more versions of this product

Catalog No. AAA1345036


Explore available promotions

Description

Description

1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

Solubility
It is Soluble in ether, chloroform, and aqueous sodium hydroxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Acid anhydrides, Halogens, halogen acids. Stable under recommended storage conditions.
Specifications

Specifications

1,3-Diphenyl-1,3-propanedione
76°C to 80°C
C15H12O2
500 g
14,3009
It is Soluble in ether,chloroform,and aqueous sodium hydroxide. Insoluble in water.
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2
224.259
CHEBI:75417
≥98%
120-46-7
219°C to 221°C (18 mmHg)
MFCD00003085
514910
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
NZZIMKJIVMHWJC-UHFFFAOYSA-N
1,3-diphenylpropane-1,3-dione
8433
224.26

Chemical Identifiers

120-46-7
224.259
NZZIMKJIVMHWJC-UHFFFAOYSA-N
8433
1,3-diphenylpropane-1,3-dione
C15H12O2
MFCD00003085
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
CHEBI:75417
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-398-9

RTECSNumber : TZ1930000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only