missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Diphenyl-1,3-propanedione, 98+%, Thermo Scientific Chemicals

$91.75 - $959.84

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00003085
InChI Key NZZIMKJIVMHWJC-UHFFFAOYSA-N
Synonym dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
PubChem CID 8433
ChEBI CHEBI:75417
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1345014
View Documents
Thermo Scientific Chemicals
A1345014
25 g
Each for $91.75
 
AAA1345022
View Documents
Thermo Scientific Chemicals
A1345022
100 g
Each for $273.82
 
AAA1345036
View Documents
Thermo Scientific Chemicals
A1345036
500 g
Each for $959.84
 
Description

Description

1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer & antitumour agent. It is used in perfumery and as well as in organic synthesis.

Solubility
It is Soluble in ether, chloroform, and aqueous sodium hydroxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Acid anhydrides, Halogens, halogen acids. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

120-46-7
224.259
NZZIMKJIVMHWJC-UHFFFAOYSA-N
8433
1,3-diphenylpropane-1,3-dione
C15H12O2
MFCD00003085
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
CHEBI:75417
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2

Specifications

120-46-7
219°C to 221°C (18 mmHg)
MFCD00003085
514910
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
NZZIMKJIVMHWJC-UHFFFAOYSA-N
1,3-diphenylpropane-1,3-dione
8433
224.26
1,3-Diphenyl-1,3-propanedione
76°C to 80°C
C15H12O2
25 g
14,3009
It is Soluble in ether,chloroform,and aqueous sodium hydroxide. Insoluble in water.
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2
224.259
CHEBI:75417
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-398-9

RTECSNumber : TZ1930000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only