missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Diphenylacetone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 117170250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 3-Diphenylacetone | |
98.5 | |
30.0°C to 34.0°C | |
1.0400g/mL | |
>110°C | |
98.5% min. (GC) | |
C15H14O | |
MFCD00004795 | |
07,445 | |
1.04 | |
Solubility in water: insoluble. Other solubilities: soluble ethanol, diethyl ether, petroleum ether | |
O=C(CC1=CC=CC=C1)CC1=CC=CC=C1 | |
210.28 | |
210.27 | |
Crystalline Mass or Liquid |
102-04-5 | |
100.0 | |
Yellow | |
330.0°C | |
Authentic | |
Glass bottle | |
C6H5CH2COCH2C6H5 | |
25 g | |
01,198; 04,123 | |
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397 | |
YFKBXYGUSOXJGS-UHFFFAOYSA-N | |
1,3-diphenylpropan-2-one | |
7593 | |
99% |
Chemical Identifiers
102-04-5 | |
210.28 | |
YFKBXYGUSOXJGS-UHFFFAOYSA-N | |
7593 | |
O=C(CC1=CC=CC=C1)CC1=CC=CC=C1 |
C15H14O | |
MFCD00004795 | |
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397 | |
1,3-diphenylpropan-2-one |
Safety and Handling
EINECSNumber : 203-000-
RUO – Research Use Only