missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Diphenylacetone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117171000

 View more versions of this product

Catalog No. AC117171000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 3-Diphenylacetone
98.5
30.0°C to 34.0°C
1.0400g/mL
>110°C
98.5% min. (GC)
C15H14O
MFCD00004795
07,445
1.04
Solubility in water: insoluble. Other solubilities: soluble ethanol, diethyl ether, petroleum ether
O=C(CC1=CC=CC=C1)CC1=CC=CC=C1
210.28
210.27
Crystalline Mass or Liquid
102-04-5
100.0
Yellow
330.0°C
Authentic
Glass bottle
C6H5CH2COCH2C6H5
100 g
01,198; 04,123
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
YFKBXYGUSOXJGS-UHFFFAOYSA-N
1,3-diphenylpropan-2-one
7593
99%

Chemical Identifiers

102-04-5
210.28
YFKBXYGUSOXJGS-UHFFFAOYSA-N
7593
O=C(CC1=CC=CC=C1)CC1=CC=CC=C1
C15H14O
MFCD00004795
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
1,3-diphenylpropan-2-one
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-000-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only