missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[4-(3-Bromothien-2-yl)phenyl]ethanone, 97%, Thermo Scientific™

Chemical Identifiers

CAS 937796-01-5
Molecular Formula C12H9BrOS
Molecular Weight (g/mol) 281.167
MDL Number MFCD09879973
InChI Key AMADTVLVNSCDKW-UHFFFAOYSA-N
Synonym 1-4-3-bromothien-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 4'-3-bromothien-2-yl acetophenone
PubChem CID 24229756
IUPAC Name 1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=C(C=CS2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC67828DA
SDS View Documents
Thermo Scientific Chemicals
CC67828DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC67828DE
SDS View Documents
Thermo Scientific Chemicals
CC67828DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

937796-01-5
281.167
AMADTVLVNSCDKW-UHFFFAOYSA-N
24229756
CC(=O)C1=CC=C(C=C1)C2=C(C=CS2)Br
C12H9BrOS
MFCD09879973
1-4-3-bromothien-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 4'-3-bromothien-2-yl acetophenone
1-[4-(3-bromothiophen-2-yl)phenyl]ethanone

Specifications

937796-01-5
100.0
Amber Glass Bottle
MFCD09879973
1-4-3-bromothien-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 4'-3-bromothien-2-yl acetophenone
CC(=O)C1=CC=C(C=C1)C2=C(C=CS2)Br
281.167
281.17
1-[4-(3-Bromothien-2-yl)phenyl]ethanone
96.5
97%
C12H9BrOS
1g
AMADTVLVNSCDKW-UHFFFAOYSA-N
1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
24229756
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'