missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Biphenylyl)ethylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5823703

 View more versions of this product

Catalog No. AAH5823703


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
Specifications

Specifications

1-(4-Biphenylyl)ethylamine
C14H15N
1 g
QVLZRPPCCDKMPO-UHFFFAOYSA-N
1-(4-phenylphenyl)ethanamine
2771754
97%
86217-82-5
MFCD02667819
1-4-phenylphenyl ethanamine, 1-1,1'-biphenyl-4-yl ethanamine, chembl19081, 1,1'-biphenyl-4-methanamine,a-methyl, 1-4-phenylphenyl ethylamine, 4-1-aminoethyl biphenyl, 1-4-biphenylyl ethylamine, 1-biphenyl-4-yl-ethylamine
CC(C1=CC=C(C=C1)C2=CC=CC=C2)N
197.281
197.28

Chemical Identifiers

86217-82-5
197.281
QVLZRPPCCDKMPO-UHFFFAOYSA-N
2771754
CC(C1=CC=C(C=C1)C2=CC=CC=C2)N
C14H15N
MFCD02667819
1-4-phenylphenyl ethanamine, 1-1,1'-biphenyl-4-yl ethanamine, chembl19081, 1,1'-biphenyl-4-methanamine,a-methyl, 1-4-phenylphenyl ethylamine, 4-1-aminoethyl biphenyl, 1-4-biphenylyl ethylamine, 1-biphenyl-4-yl-ethylamine
1-(4-phenylphenyl)ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions