missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(4-Biphenylyl)ethylamine, 97%, Thermo Scientific™
$407.99 - $1600.85
Chemical Identifiers
CAS | 86217-82-5 |
---|---|
Molecular Formula | C14H15N |
Molecular Weight (g/mol) | 197.281 |
MDL Number | MFCD02667819 |
InChI Key | QVLZRPPCCDKMPO-UHFFFAOYSA-N |
Synonym | 1-4-phenylphenyl ethanamine, 1-1,1'-biphenyl-4-yl ethanamine, chembl19081, 1,1'-biphenyl-4-methanamine,a-methyl, 1-4-phenylphenyl ethylamine, 4-1-aminoethyl biphenyl, 1-4-biphenylyl ethylamine, 1-biphenyl-4-yl-ethylamine |
PubChem CID | 2771754 |
IUPAC Name | 1-(4-phenylphenyl)ethanamine |
SMILES | CC(C1=CC=C(C=C1)C2=CC=CC=C2)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5823703
|
Thermo Scientific Chemicals
H5823703 |
1 g |
Each for $407.99
|
|
|||||
AAH5823706
|
Thermo Scientific Chemicals
H5823706 |
5 g |
Each for $1,600.85
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as pharmaceutical intermediate.
Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
Chemical Identifiers
86217-82-5 | |
197.281 | |
QVLZRPPCCDKMPO-UHFFFAOYSA-N | |
2771754 | |
CC(C1=CC=C(C=C1)C2=CC=CC=C2)N |
C14H15N | |
MFCD02667819 | |
1-4-phenylphenyl ethanamine, 1-1,1'-biphenyl-4-yl ethanamine, chembl19081, 1,1'-biphenyl-4-methanamine,a-methyl, 1-4-phenylphenyl ethylamine, 4-1-aminoethyl biphenyl, 1-4-biphenylyl ethylamine, 1-biphenyl-4-yl-ethylamine | |
1-(4-phenylphenyl)ethanamine |
Specifications
86217-82-5 | |
MFCD02667819 | |
1-4-phenylphenyl ethanamine, 1-1,1'-biphenyl-4-yl ethanamine, chembl19081, 1,1'-biphenyl-4-methanamine,a-methyl, 1-4-phenylphenyl ethylamine, 4-1-aminoethyl biphenyl, 1-4-biphenylyl ethylamine, 1-biphenyl-4-yl-ethylamine | |
CC(C1=CC=C(C=C1)C2=CC=CC=C2)N | |
197.281 | |
197.28 | |
1-(4-Biphenylyl)ethylamine |
C14H15N | |
1 g | |
QVLZRPPCCDKMPO-UHFFFAOYSA-N | |
1-(4-phenylphenyl)ethanamine | |
2771754 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures