missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-1-(4-Bromophenyl)ethylamine, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1184806

 View more versions of this product

Catalog No. AAL1184806


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+/-)-1-(4-Bromophenyl)ethylamine
1.397
>110°C (230°F)
C8H10BrN
MFCD00025548
UN2735
Air Sensitive
SOZMSEPDYJGBEK-UHFFFAOYSA-N
1-(4-bromophenyl)ethanamine
91175
96%
24358-62-1
140°C to 145°C (30 mmHg)
Amine-like
1.567
5 g
2412318
1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
CC(C1=CC=C(C=C1)Br)N
200.079
200.08

Chemical Identifiers

24358-62-1
200.079
SOZMSEPDYJGBEK-UHFFFAOYSA-N
91175
CC(C1=CC=C(C=C1)Br)N
C8H10BrN
MFCD00025548
1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
1-(4-bromophenyl)ethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 246-200-3

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only