missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-1-(4-Bromophenyl)ethylamine, 96%, Thermo Scientific Chemicals

$47.11 - $106.66

Chemical Identifiers

CAS 24358-62-1
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00025548
InChI Key SOZMSEPDYJGBEK-UHFFFAOYSA-N
Synonym 1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
PubChem CID 91175
IUPAC Name 1-(4-bromophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1184803
View Documents
Thermo Scientific Chemicals
L1184803
1 g
Each for $47.11
 
AAL1184806
View Documents
Thermo Scientific Chemicals
L1184806
5 g
Each for $106.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24358-62-1
200.079
SOZMSEPDYJGBEK-UHFFFAOYSA-N
91175
CC(C1=CC=C(C=C1)Br)N
C8H10BrN
MFCD00025548
1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
1-(4-bromophenyl)ethanamine

Specifications

24358-62-1
140°C to 145°C (30 mmHg)
Amine-like
1.567
1 g
2412318
1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
CC(C1=CC=C(C=C1)Br)N
200.079
200.08
(+/-)-1-(4-Bromophenyl)ethylamine
1.397
>110°C (230°F)
C8H10BrN
MFCD00025548
UN2735
Air Sensitive
SOZMSEPDYJGBEK-UHFFFAOYSA-N
1-(4-bromophenyl)ethanamine
91175
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 246-200-3

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only