missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Bromophenyl)pyrrolidin-2-one, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5056303

 View more versions of this product

Catalog No. AAH5056303

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-(4-Bromophenyl)pyrrolidin-2-one
98°C to 101°C
MFCD00138502
1-4-bromophenyl pyrrolidin-2-one, 1-4-bromophenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-4-bromophenyl, 1-4-bromophenyl-2-pyrrolidone, pubchem9356, p-bromo-phenylpyrrolidone, acmc-209p4u, buttpark 32\07-61, n-4-bromophenyl-2-pyrrolidinone, 1-4-bromophenyl pyrrolidine-2-one
C1CC(=O)N(C1)C2=CC=C(C=C2)Br
240.1
240.1
7661-32-7
C10H10BrNO
1 g
YINFEFUSAQRZGG-UHFFFAOYSA-N
1-(4-bromophenyl)pyrrolidin-2-one
736110
98%

Chemical Identifiers

7661-32-7
240.1
YINFEFUSAQRZGG-UHFFFAOYSA-N
736110
C1CC(=O)N(C1)C2=CC=C(C=C2)Br
C10H10BrNO
MFCD00138502
1-4-bromophenyl pyrrolidin-2-one, 1-4-bromophenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-4-bromophenyl, 1-4-bromophenyl-2-pyrrolidone, pubchem9356, p-bromo-phenylpyrrolidone, acmc-209p4u, buttpark 32\07-61, n-4-bromophenyl-2-pyrrolidinone, 1-4-bromophenyl pyrrolidine-2-one
1-(4-bromophenyl)pyrrolidin-2-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only