missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Bromophenyl)pyrrolidin-2-one, 98%, Thermo Scientific Chemicals

$195.40 - $832.74

Chemical Identifiers

CAS 7661-32-7
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD00138502
InChI Key YINFEFUSAQRZGG-UHFFFAOYSA-N
Synonym 1-4-bromophenyl pyrrolidin-2-one, 1-4-bromophenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-4-bromophenyl, 1-4-bromophenyl-2-pyrrolidone, pubchem9356, p-bromo-phenylpyrrolidone, acmc-209p4u, buttpark 32\07-61, n-4-bromophenyl-2-pyrrolidinone, 1-4-bromophenyl pyrrolidine-2-one
PubChem CID 736110
IUPAC Name 1-(4-bromophenyl)pyrrolidin-2-one
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5056303
View Documents
Thermo Scientific Chemicals
H5056303
1 g
Each for $195.40
 
AAH5056306
View Documents
Thermo Scientific Chemicals
H5056306
5 g
Each for $832.74
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7661-32-7
240.1
YINFEFUSAQRZGG-UHFFFAOYSA-N
736110
C1CC(=O)N(C1)C2=CC=C(C=C2)Br
C10H10BrNO
MFCD00138502
1-4-bromophenyl pyrrolidin-2-one, 1-4-bromophenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-4-bromophenyl, 1-4-bromophenyl-2-pyrrolidone, pubchem9356, p-bromo-phenylpyrrolidone, acmc-209p4u, buttpark 32\07-61, n-4-bromophenyl-2-pyrrolidinone, 1-4-bromophenyl pyrrolidine-2-one
1-(4-bromophenyl)pyrrolidin-2-one

Specifications

7661-32-7
C10H10BrNO
1 g
YINFEFUSAQRZGG-UHFFFAOYSA-N
1-(4-bromophenyl)pyrrolidin-2-one
736110
98%
98°C to 101°C
MFCD00138502
1-4-bromophenyl pyrrolidin-2-one, 1-4-bromophenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-4-bromophenyl, 1-4-bromophenyl-2-pyrrolidone, pubchem9356, p-bromo-phenylpyrrolidone, acmc-209p4u, buttpark 32\07-61, n-4-bromophenyl-2-pyrrolidinone, 1-4-bromophenyl pyrrolidine-2-one
C1CC(=O)N(C1)C2=CC=C(C=C2)Br
240.1
240.1
1-(4-Bromophenyl)pyrrolidin-2-one
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only