missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 161570050

 View more versions of this product

Catalog No. AC161570050


Explore available promotions

Description

Description

Specifications

Specifications

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid
80789-69-1
97.5
5 g
Glass bottle
98%
ClC6H4C5H8CO2H
1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid
C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O
224.69
224.69
160.0°C to 164.0°C
Cream to Gray
100.0
Authentic
Crystalline Powder or Crystals
C12H13ClO2
MFCD00001373
QJNFJEMGWIQMJT-UHFFFAOYSA-N
1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
97447
98%

Chemical Identifiers

80789-69-1
224.69
QJNFJEMGWIQMJT-UHFFFAOYSA-N
97447
C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O
C12H13ClO2
MFCD00001373
1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid
1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 279-552-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only