missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Cyclohexadiene, 97%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 154820050

 View more versions of this product

Catalog No. AC154820050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 4-Cyclohexadiene
628-41-1
100.0
Colorless
81.0°C to 82.0°C
Authentic
Glass bottle
1.4710 to 1.4730
5 mL
1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
C1C=CCC=C1
80.13
12343
80.13
Liquid
Stabilized
96.0
-49.2°C
0.8400g/mL
−6°C
96% min. (GC)
C6H8
MFCD00001535
0.84
UVJHQYIOXKWHFD-UHFFFAOYSA-N
cyclohexa-1,4-diene
0.1% BHT
CHEBI:37611
97%

Chemical Identifiers

628-41-1
80.13
UVJHQYIOXKWHFD-UHFFFAOYSA-N
12343
cyclohexa-1,4-diene
C6H8
MFCD00001535
1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
CHEBI:37611
C1C=CCC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-043-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only