Learn More
1,4-Cyclohexadiene, 97%, stabilized, Thermo Scientific Chemicals
$88.21 - $996.39
Chemical Identifiers
CAS | 628-41-1 |
---|---|
Molecular Formula | C6H8 |
Molecular Weight (g/mol) | 80.13 |
MDL Number | MFCD00001535 |
InChI Key | UVJHQYIOXKWHFD-UHFFFAOYSA-N |
Synonym | 1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc |
PubChem CID | 12343 |
ChEBI | CHEBI:37611 |
IUPAC Name | cyclohexa-1,4-diene |
SMILES | C1C=CCC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC154820050
|
Thermo Scientific Chemicals
154820050 |
5 mL | Glass bottle |
Each for $88.21
|
|
||||
AC154820250
|
Thermo Scientific Chemicals
154820250 |
25 mL | Glass bottle |
Each for $275.81
|
|
||||
AC154821000
|
Thermo Scientific Chemicals
154821000 |
100 mL | Glass bottle |
Each for $996.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
628-41-1 | |
80.13 | |
UVJHQYIOXKWHFD-UHFFFAOYSA-N | |
12343 | |
cyclohexa-1,4-diene |
C6H8 | |
MFCD00001535 | |
1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc | |
CHEBI:37611 | |
C1C=CCC=C1 |
Specifications
628-41-1 | |
100.0 | |
Colorless | |
81.0°C to 82.0°C | |
Authentic | |
Glass bottle | |
1.4710 to 1.4730 | |
5 mL | |
1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc | |
C1C=CCC=C1 | |
80.13 | |
12343 | |
80.13 | |
Liquid |
96.0 | |
-49.2°C | |
0.8400g/mL | |
−6°C | |
96% min. (GC) | |
C6H8 | |
MFCD00001535 | |
0.84 | |
UVJHQYIOXKWHFD-UHFFFAOYSA-N | |
cyclohexa-1,4-diene | |
0.1% BHT | |
CHEBI:37611 | |
97% | |
1, 4-Cyclohexadiene, Stabilized |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 211-043-1
RUO – Research Use Only