Learn More
1,4-Diaminobutane dihydrochloride, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 215111000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 4-Diaminobutane dihydrochloride | |
99.0 | |
280.0°C | |
Authentic | |
Glass bottle | |
H2N(CH2)4NH2·2HCl | |
100 g | |
15, 8058 | |
Solubility in water: soluble. | |
C(CCN)CN.Cl.Cl | |
161.08 | |
161.08 | |
Crystalline Powder |
333-93-7 | |
100.0 | |
White | |
99+% | |
C4H12N2·2HCl | |
MFCD00012526 | |
04, 264 | |
1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride | |
XXWCODXIQWIHQN-UHFFFAOYSA-N | |
butane-1,4-diamine;dihydrochloride | |
9532 | |
99+% |
Chemical Identifiers
333-93-7 | |
161.08 | |
XXWCODXIQWIHQN-UHFFFAOYSA-N | |
9532 | |
C(CCN)CN.Cl.Cl |
C4H12N2·2HCl | |
MFCD00012526 | |
1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride | |
butane-1,4-diamine;dihydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious
GHS Signal Word: Warning
EINECSNumber : 206-375-9
RUO – Research Use Only