missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Diaminobutane dihydrochloride, 99+%, Thermo Scientific Chemicals

$43.55 - $362.61

Chemical Identifiers

CAS 333-93-7
Molecular Formula C4H12N2·2HCl
Molecular Weight (g/mol) 161.08
MDL Number MFCD00012526
InChI Key XXWCODXIQWIHQN-UHFFFAOYSA-N
Synonym 1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
PubChem CID 9532
IUPAC Name butane-1,4-diamine;dihydrochloride
SMILES C(CCN)CN.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC215110050
SDS View Documents
Thermo Scientific Chemicals
215110050
5 g Glass bottle
Each for $43.55
 
AC215111000
SDS View Documents
Thermo Scientific Chemicals
215111000
100 g Glass bottle
Each for $362.61
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

333-93-7
161.08
XXWCODXIQWIHQN-UHFFFAOYSA-N
9532
C(CCN)CN.Cl.Cl
C4H12N2·2HCl
MFCD00012526
1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
butane-1,4-diamine;dihydrochloride

Specifications

333-93-7
100.0
White
99+%
C4H12N2·2HCl
MFCD00012526
04, 264
1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
XXWCODXIQWIHQN-UHFFFAOYSA-N
butane-1,4-diamine;dihydrochloride
9532
99+%
1, 4-Diaminobutane dihydrochloride
99.0
280.0°C
Authentic
Glass bottle
H2N(CH2)4NH2·2HCl
5 g
15, 8058
Solubility in water: soluble.
C(CCN)CN.Cl.Cl
161.08
161.08
Crystalline Powder
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious

GHS Signal Word: Warning

EINECSNumber : 206-375-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only