missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 113190025

 View more versions of this product

Catalog No. AC113190025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 4-Dichlorobenzene
106-46-7
100.0
52°C to 56°C
1.2400g/mL
67°C
99% min. (GC)
C6H4Cl2
MFCD00000604
05, 203
15, 3066
Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether,chloroform,and carbon disulfide
C1=CC(=CC=C1Cl)Cl
147
CHEBI:28618
1.7mbar at 20°C
Crystals
97%
99.0
>173°C
White
174°C
Authentic
Plastic bottle
C6H4Cl2
2.5 kg
1.24
p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
OCJBOOLMMGQPQU-UHFFFAOYSA-N
1,4-dichlorobenzene
4685
147
99+%

Chemical Identifiers

106-46-7
147
OCJBOOLMMGQPQU-UHFFFAOYSA-N
4685
1,4-dichlorobenzene
C6H4Cl2
MFCD00000604
p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
CHEBI:28618
C1=CC(=CC=C1Cl)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Suspected of causing cancer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Warning

EINECSNumber : 203-400-5

RTECSNumber : CZ4550000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only