missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobenzene, 99+%

CAS: 106-46-7 | C6H4Cl2 | 147 g/mol

$103.75 - $186.22

Chemical Identifiers

CAS 106-46-7
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 147
MDL Number MFCD00000604
InChI Key OCJBOOLMMGQPQU-UHFFFAOYSA-N
Synonym p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
PubChem CID 4685
ChEBI CHEBI:28618
IUPAC Name 1,4-dichlorobenzene
SMILES C1=CC(=CC=C1Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC113190010
SDS View Documents
Thermo Scientific Chemicals
113190010
1 kg Plastic bottle
Each for $103.75
Only null left
 
AC113190025
SDS View Documents
Thermo Scientific Chemicals
113190025
2.5 kg Plastic bottle
Each for $186.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-46-7
147
OCJBOOLMMGQPQU-UHFFFAOYSA-N
4685
1,4-dichlorobenzene
C6H4Cl2
MFCD00000604
p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
CHEBI:28618
C1=CC(=CC=C1Cl)Cl

Specifications

106-46-7
100.0
52°C to 56°C
1.2400g/mL
67°C
99% min. (GC)
C6H4Cl2
MFCD00000604
05, 203
15, 3066
Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether,chloroform,and carbon disulfide
C1=CC(=CC=C1Cl)Cl
147
CHEBI:28618
1.7mbar at 20°C
Crystals
99.0
>173°C
White
174°C
Authentic
Plastic bottle
C6H4Cl2
1 kg
1.24
p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
OCJBOOLMMGQPQU-UHFFFAOYSA-N
1,4-dichlorobenzene
4685
147
99+%
1, 4-Dichlorobenzene, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Suspected of causing cancer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Warning

EINECSNumber : 203-400-5

RTECSNumber : CZ4550000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only