missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 114150050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 4-Diethylbenzene | |
105-05-5 | |
100.0 | |
Colorless | |
184°C | |
Authentic | |
Glass bottle | |
1.4935 to 1.4955 | |
MFCD00009264 | |
05, 426 | |
p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711 | |
DSNHSQKRULAAEI-UHFFFAOYSA-N | |
1,4-diethylbenzene | |
7734 | |
134.22 | |
Liquid |
98% | |
97.5 | |
-43°C | |
0.8600g/mL | |
56°C | |
97.5% min. (GC) | |
C10H14 | |
C6H4(C2H5)2 | |
5 mL | |
0.86 | |
Solubility in water: insoluble in water. Other solubilities: soluble in benzene and carbon tetrachloride,miscible with ethanol,ether and acetone | |
CCC1=CC=C(CC)C=C1 | |
134.22 | |
CHEBI:34062 | |
98% |
Chemical Identifiers
105-05-5 | |
134.22 | |
DSNHSQKRULAAEI-UHFFFAOYSA-N | |
7734 | |
1,4-diethylbenzene |
C10H14 | |
MFCD00009264 | |
p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711 | |
CHEBI:34062 | |
CCC1=CC=C(CC)C=C1 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 203-265-2
TSCA : TSCA
RUO – Research Use Only