missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Hexadiene (cis- and trans- mixture) 99.0+%, TCI America™
$316.00 - $316.00
Chemical Identifiers
CAS | 592-45-0 |
---|---|
Molecular Formula | C6H10 |
Molecular Weight (g/mol) | 82.146 |
MDL Number | MFCD00009298 |
InChI Key | PRBHEGAFLDMLAL-GQCTYLIASA-N |
Synonym | 1,4-hexadiene, trans-1,4-hexadiene, 1,4-hexadiene, 4e, 1,4-hexadiene,c&t, allylpropenyl, 1,4-hexadiene cis-trans, 1,4-hexa-1,4-diene, 1-allylpropene, 1,4-hexadiene,cis-trans, 1,4-trans-hexadiene |
PubChem CID | 5365552 |
IUPAC Name | (4E)-hexa-1,4-diene |
SMILES | CC=CCC=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
H03335ML
|
TCI America
H03335ML |
5 mL |
Each for $316.00
|
|
||||||
Chemical Identifiers
592-45-0 | |
82.146 | |
PRBHEGAFLDMLAL-GQCTYLIASA-N | |
5365552 | |
CC=CCC=C |
C6H10 | |
MFCD00009298 | |
1,4-hexadiene, trans-1,4-hexadiene, 1,4-hexadiene, 4e, 1,4-hexadiene,c&t, allylpropenyl, 1,4-hexadiene cis-trans, 1,4-hexa-1,4-diene, 1-allylpropene, 1,4-hexadiene,cis-trans, 1,4-trans-hexadiene | |
(4E)-hexa-1,4-diene |
Specifications
592-45-0 | |
65°C | |
MFCD00009298 | |
2458 | |
PRBHEGAFLDMLAL-GQCTYLIASA-N | |
(4E)-hexa-1,4-diene | |
5365552 | |
≥99.0% (GC) | |
1,4-Hexadiene (cis- and trans- mixture) |
Yellow | |
C6H10 | |
5 mL | |
1,4-hexadiene, trans-1,4-hexadiene, 1,4-hexadiene, 4e, 1,4-hexadiene,c&t, allylpropenyl, 1,4-hexadiene cis-trans, 1,4-hexa-1,4-diene, 1-allylpropene, 1,4-hexadiene,cis-trans, 1,4-trans-hexadiene | |
CC=CCC=C | |
82.146 | |
82.15 | |
Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator