missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Naphthalenedicarboxylic Acid 95.0+%, TCI America™
Supplier: TCI America N060625G
Specifications
1,4-Naphthalenedicarboxylic Acid | |
320°C | |
MFCD00014312 | |
1,4-naphthalenedicarboxylic acid, 1,4-naphthalenedicarboxylicacid, 1,4-naphthalicacid, 1,4-naphthalic acid, pubchem21317, acmc-1b0c7, ksc357a6b, naphthalene-1,4-dicarboxylicacid, naphthalene-1,4-dicarboxlic acid, naphthaline-1,4-dicarboxylic acid | |
OC(=O)C1=C2C=CC=CC2=C(C=C1)C(O)=O | |
216.19 | |
216.19 | |
Crystalline Powder |
605-70-9 | |
C12H8O4 | |
25 g | |
ABMFBCRYHDZLRD-UHFFFAOYSA-N | |
naphthalene-1,4-dicarboxylic acid | |
69065 | |
≥95.0% (T) |
Chemical Identifiers
605-70-9 | |
216.19 | |
ABMFBCRYHDZLRD-UHFFFAOYSA-N | |
69065 | |
OC(=O)C1=C2C=CC=CC2=C(C=C1)C(O)=O |
C12H8O4 | |
MFCD00014312 | |
1,4-naphthalenedicarboxylic acid, 1,4-naphthalenedicarboxylicacid, 1,4-naphthalicacid, 1,4-naphthalic acid, pubchem21317, acmc-1b0c7, ksc357a6b, naphthalene-1,4-dicarboxylicacid, naphthalene-1,4-dicarboxlic acid, naphthaline-1,4-dicarboxylic acid | |
naphthalene-1,4-dicarboxylic acid |
Safety and Handling
EINECSNumber : (4)-1765
TSCA : Yes