missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Phenylene Diisocyanate 98.0+%, TCI America™
$116.52 - $566.83
Chemical Identifiers
CAS | 104-49-4 |
---|---|
Molecular Formula | C8H4N2O2 |
Molecular Weight (g/mol) | 160.132 |
InChI Key | ALQLPWJFHRMHIU-UHFFFAOYSA-N |
Synonym | Benzene 1,4-Diisocyanate |
PubChem CID | 61009 |
IUPAC Name | 1,4-diisocyanatobenzene |
SMILES | C1=CC(=CC=C1N=C=O)N=C=O |
Chemical Identifiers
104-49-4 | |
160.132 | |
Benzene 1,4-Diisocyanate | |
1,4-diisocyanatobenzene |
C8H4N2O2 | |
ALQLPWJFHRMHIU-UHFFFAOYSA-N | |
61009 | |
C1=CC(=CC=C1N=C=O)N=C=O |
Specifications
104-49-4 | |
White-Yellow | |
C8H4N2O2 | |
2811 | |
ALQLPWJFHRMHIU-UHFFFAOYSA-N | |
1,4-diisocyanatobenzene | |
61009 | |
≥98.0% (GC) | |
1,4-Phenylene Diisocyanate |
96°C | |
118°C | |
25 g | |
Benzene 1,4-Diisocyanate | |
C1=CC(=CC=C1N=C=O)N=C=O | |
160.132 | |
160.13 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2215
RTECSNumber : CZ6150000
TSCA : Yes
Recommended Storage : Refrigerator