missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Phenylene Diisocyanate 98.0+%, TCI America™

Supplier:  TCI America P164025G

 View more versions of this product

Catalog No. P164025G


Explore available promotions

Description

Description

Specifications

Specifications

1,4-Phenylene Diisocyanate
96°C
118°C
25 g
Benzene 1,4-Diisocyanate
C1=CC(=CC=C1N=C=O)N=C=O
160.132
160.13
Crystalline Powder
104-49-4
White-Yellow
C8H4N2O2
2811
ALQLPWJFHRMHIU-UHFFFAOYSA-N
1,4-diisocyanatobenzene
61009
≥98.0% (GC)

Chemical Identifiers

104-49-4
160.132
Benzene 1,4-Diisocyanate
1,4-diisocyanatobenzene
C8H4N2O2
ALQLPWJFHRMHIU-UHFFFAOYSA-N
61009
C1=CC(=CC=C1N=C=O)N=C=O
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2215

RTECSNumber : CZ6150000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions