missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,5-Dibromo-3-methylpentane, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2052114

 View more versions of this product

Catalog No. AAB2052114



Description

Description

1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
Specifications

Specifications

1,5-Dibromo-3-methylpentane
1.58
101°C (214°F)
BrCH2CH2CH(CH3)CH2CH2Br
MFCD00039200
3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g
YDPZWUMQKMLLHC-UHFFFAOYSA-N
1,5-dibromo-3-methylpentane
138258
≥98%
4457-72-1
233°C
C6H12Br2
1.508
25 g
Insoluble in water.
CC(CCBr)CCBr
243.97
243.98

Chemical Identifiers

4457-72-1
243.97
YDPZWUMQKMLLHC-UHFFFAOYSA-N
138258
CC(CCBr)CCBr
C6H12Br2
MFCD00039200
3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g
1,5-dibromo-3-methylpentane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only