Learn More
1,5-Dibromo-3-methylpentane, 98+%, Thermo Scientific Chemicals
$110.74 - $375.19
Chemical Identifiers
CAS | 4457-72-1 |
---|---|
Molecular Formula | C6H12Br2 |
Molecular Weight (g/mol) | 243.97 |
MDL Number | MFCD00039200 |
InChI Key | YDPZWUMQKMLLHC-UHFFFAOYSA-N |
Synonym | 3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g |
PubChem CID | 138258 |
IUPAC Name | 1,5-dibromo-3-methylpentane |
SMILES | CC(CCBr)CCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2052106
|
Thermo Scientific Chemicals
B2052106 |
5 g |
Each for $110.74
|
|
|||||
AAB2052114
|
Thermo Scientific Chemicals
B2052114 |
25 g |
Each for $375.19
|
|
|||||
Description
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
Chemical Identifiers
4457-72-1 | |
243.97 | |
YDPZWUMQKMLLHC-UHFFFAOYSA-N | |
138258 | |
CC(CCBr)CCBr |
C6H12Br2 | |
MFCD00039200 | |
3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g | |
1,5-dibromo-3-methylpentane |
Specifications
4457-72-1 | |
233°C | |
C6H12Br2 | |
1.508 | |
5 g | |
Insoluble in water. | |
CC(CCBr)CCBr | |
243.97 | |
243.98 | |
1,5-Dibromo-3-methylpentane |
1.58 | |
101°C (214°F) | |
BrCH2CH2CH(CH3)CH2CH2Br | |
MFCD00039200 | |
3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g | |
YDPZWUMQKMLLHC-UHFFFAOYSA-N | |
1,5-dibromo-3-methylpentane | |
138258 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only