missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Diphenyl-1,3,5-hexatriene, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117300010

 View more versions of this product

Catalog No. AC117300010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,6-Diphenyl-1,3,5-hexatriene
1720-32-7
100.0
Yellow
97.5% min. (HPLC)
C18H16
MFCD00004793
05, 691
BOBLSBAZCVBABY-WPWUJOAOSA-N
[(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
5376733
232.32
Crystalline Powder or Crystals
98%
97.5
199°C to 203°C
Authentic
Glass bottle
C6H5CH=CHCH=CHCH=CHC6H5
1 g
1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2
232.32
CHEBI:51594
98%

Chemical Identifiers

1720-32-7
232.32
BOBLSBAZCVBABY-WPWUJOAOSA-N
5376733
[(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
C18H16
MFCD00004793
1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
CHEBI:51594
C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-011-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only