missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Diphenyl-1,3,5-hexatriene, 95%, Thermo Scientific Chemicals

$120.38 - $120.38

Chemical Identifiers

CAS 1720-32-7
Molecular Formula C18H16
Molecular Weight (g/mol) 232.32
MDL Number MFCD00004793
InChI Key BOBLSBAZCVBABY-WPWUJOAOSA-N
Synonym 1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
PubChem CID 5376733
ChEBI CHEBI:51594
IUPAC Name [(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
SMILES C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC117300010
SDS View Documents
Thermo Scientific Chemicals
117300010
1 g Glass bottle
Each for $120.38
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1720-32-7
232.32
BOBLSBAZCVBABY-WPWUJOAOSA-N
5376733
[(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
C18H16
MFCD00004793
1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
CHEBI:51594
C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2

Specifications

1720-32-7
100.0
Yellow
97.5% min. (HPLC)
C18H16
MFCD00004793
05, 691
BOBLSBAZCVBABY-WPWUJOAOSA-N
[(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
5376733
232.32
Crystalline Powder or Crystals
97.5
199°C to 203°C
Authentic
Glass bottle
C6H5CH=CHCH=CHCH=CHC6H5
1 g
1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2
232.32
CHEBI:51594
98%
1,6-Diphenyl-1,3,5-hexatriene, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-011-3

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only