missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(6-Methoxy-2-naphthyl)ethanol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2562806

 View more versions of this product

Catalog No. AAB2562806



Description

Description

Racemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Racemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.

Solubility
Soluble in DMSO, acetone, dichloromethane, ethylacetate. Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

1-(6-Methoxy-2-naphthyl)ethanol
111°C to 112°C
MFCD01632581
1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k
OUVJWFRUESFCCY-SECBINFHSA-N
(1R)-1-(6-methoxynaphthalen-2-yl)ethan-1-ol
575523
98%
77301-42-9
C13H14O2
5 g
Soluble in DMSO,acetone,dichloromethane,ethylacetate. Insoluble in water.
COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O
202.25
202.25

Chemical Identifiers

77301-42-9
202.25
OUVJWFRUESFCCY-SECBINFHSA-N
575523
C13H14O2
MFCD01632581
1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k
COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only