missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(6-Methoxy-2-naphthyl)ethanol, 98%, Thermo Scientific Chemicals
$116.34 - $340.62
Chemical Identifiers
CAS | 77301-42-9 |
---|---|
Molecular Formula | C13H14O2 |
Molecular Weight (g/mol) | 202.25 |
MDL Number | MFCD01632581 |
InChI Key | OUVJWFRUESFCCY-SECBINFHSA-N |
Synonym | 1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k |
PubChem CID | 575523 |
SMILES | COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2562806
|
Thermo Scientific Chemicals
B2562806 |
5 g |
Each for $116.34
|
|
|||||
AAB2562814
|
Thermo Scientific Chemicals
B2562814 |
25 g |
Each for $340.62
|
|
|||||
Description
Racemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRacemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.
Solubility
Soluble in DMSO, acetone, dichloromethane, ethylacetate. Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
77301-42-9 | |
202.25 | |
OUVJWFRUESFCCY-SECBINFHSA-N | |
575523 |
C13H14O2 | |
MFCD01632581 | |
1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k | |
COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O |
Specifications
77301-42-9 | |
C13H14O2 | |
5 g | |
Soluble in DMSO,acetone,dichloromethane,ethylacetate. Insoluble in water. | |
COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O | |
202.25 | |
202.25 | |
1-(6-Methoxy-2-naphthyl)ethanol |
111°C to 112°C | |
MFCD01632581 | |
1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k | |
OUVJWFRUESFCCY-SECBINFHSA-N | |
(1R)-1-(6-methoxynaphthalen-2-yl)ethan-1-ol | |
575523 | |
98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only