missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,8-Diaminooctane, 98%, Thermo Scientific Chemicals

$83.18 - $205.87

Chemical Identifiers

CAS 373-44-4
Molecular Formula C8H22Cl2N2
Molecular Weight (g/mol) 217.18
MDL Number MFCD00008248
InChI Key ZFLWZOGXFQNIMT-UHFFFAOYSA-N
Synonym 1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
PubChem CID 24250
ChEBI CHEBI:73112
SMILES [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC112310250
SDS View Documents
Thermo Scientific Chemicals
112310250
25 g Glass bottle
Each for $83.18
 
AC112311000
SDS View Documents
Thermo Scientific Chemicals
112311000
100 g Glass bottle
Each for $205.87
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

373-44-4
217.18
ZFLWZOGXFQNIMT-UHFFFAOYSA-N
24250
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
C8H22Cl2N2
MFCD00008248
1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
CHEBI:73112

Specifications

373-44-4
100.0
White to Yellow
225.0°C to 226.0°C
Authentic
Glass bottle
H2N(CH2)8NH2
25 g
1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
ZFLWZOGXFQNIMT-UHFFFAOYSA-N
octane-1,8-bis(aminium) dichloride
24250
144.26
Solidified Crystalline Mass
97.5
50.0°C to 54.0°C
165g/mL
115°C
97.5% min. (GC)
C8H22Cl2N2
MFCD00008248
04, 271
Solubility in water: 575g/L (20°C).
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
217.18
CHEBI:73112
98%
1, 8-Diaminooctane
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physician.
Wear

GHS Signal Word: Danger

EINECSNumber : 206-764-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only