Learn More
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 160610250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,8-Diazabicyclo[5.4.0]undec-7-ene | |
6674-22-2 | |
100.0 | |
Colorless to Yellow | |
261.0°C | |
Authentic | |
Glass bottle | |
1.5210 to 1.5230 | |
25 g | |
02,101; 04,119; 05,177; 06,158; 07,87; 11,155; 13,91; 14,109; 15,109; 16,105; 17,99 | |
15, 2996 | |
Solubility in water: soluble. | |
C1CCC2=NCCCN2CC1 | |
152.24 | |
11.76 mPa.s (20°C) | |
98+% |
≥98% | |
97.5 | |
-70.0°C | |
1.0190g/mL | |
116°C | |
98% min. (GC) | |
C9H16N2 | |
MFCD00006930 | |
23, V,5, 271 | |
1.019 | |
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene | |
GQHTUMJGOHRCHB-UHFFFAOYSA-N | |
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine | |
81184 | |
152.24 | |
Liquid |
Chemical Identifiers
6674-22-2 | |
GQHTUMJGOHRCHB-UHFFFAOYSA-N | |
81184 | |
C1CCC2=NCCCN2CC1 |
MFCD00006930 | |
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene | |
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 229-713-7
TSCA : TSCA
RUO – Research Use Only