missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals

$63.77 - $480.53

Chemical Identifiers

CAS 6674-22-2
MDL Number MFCD00006930
InChI Key GQHTUMJGOHRCHB-UHFFFAOYSA-N
Synonym 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
PubChem CID 81184
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCC2=NCCCN2CC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC160610250
SDS View Documents
Thermo Scientific Chemicals
160610250
25 g Glass bottle
Each for $63.77
 
AC160611000
SDS View Documents
Thermo Scientific Chemicals
160611000
100 g Glass bottle
Each for $151.00
 
AC160615000
SDS View Documents
Thermo Scientific Chemicals
160615000
500 g Glass bottle
Each for $480.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6674-22-2
GQHTUMJGOHRCHB-UHFFFAOYSA-N
81184
C1CCC2=NCCCN2CC1
MFCD00006930
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine

Specifications

6674-22-2
100.0
Colorless to Yellow
261.0°C
Authentic
Glass bottle
1.5210 to 1.5230
25 g
02,101; 04,119; 05,177; 06,158; 07,87; 11,155; 13,91; 14,109; 15,109; 16,105; 17,99
15, 2996
Solubility in water: soluble.
C1CCC2=NCCCN2CC1
152.24
11.76 mPa.s (20°C)
98+%
1,8-Diazabicyclo[5.4.0]undec-7-ene, ≥98%
97.5
-70.0°C
1.0190g/mL
116°C
98% min. (GC)
C9H16N2
MFCD00006930
23, V,5, 271
1.019
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
GQHTUMJGOHRCHB-UHFFFAOYSA-N
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
81184
152.24
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 229-713-7

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only