missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

1-Amino-1-cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 103470050

 Afficher plus de versions de ce produit

Numéro de catalogue. AC103470050


Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

1-Amino-1-cyclopentanecarboxylic acid
52-52-8
100.0
White to Beige
97%
C6H11NO2
MFCD00001381
14, I, 526
cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
NILQLFBWTXNUOE-UHFFFAOYSA-N
1-aminocyclopentane-1-carboxylic acid
2901
129.16
Crystalline Flakes or Powder
97%
96.0
320°C to 322°C
Authentic
Glass bottle
H2NC5H8CO2H
5 g
15, 2727
Solubility in water: ca 5g/100mL
C1CCC(C1)(C(=O)O)N
129.16
CHEBI:40547
97%

Identifiants chimiques

52-52-8
NILQLFBWTXNUOE-UHFFFAOYSA-N
2901
1-aminocyclopentane-1-carboxylic acid
MFCD00001381
cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
CHEBI:40547
C1CCC(C1)(C(=O)O)N
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-144-6

missing translation for 'rtecsNumber' : GY2625000

missing translation for 'tsca' : TSCA

FDS Documents

Documents

Offres spéciales

Offres spéciales

RUO – Research Use Only