missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Amino-1-cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals

$185.71 - $185.71

Chemical Identifiers

CAS 52-52-8
MDL Number MFCD00001381
InChI Key NILQLFBWTXNUOE-UHFFFAOYSA-N
Synonym cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
PubChem CID 2901
ChEBI CHEBI:40547
IUPAC Name 1-aminocyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC103470050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
103470050
5 g Glass bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52-52-8
NILQLFBWTXNUOE-UHFFFAOYSA-N
2901
1-aminocyclopentane-1-carboxylic acid
MFCD00001381
cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
CHEBI:40547
C1CCC(C1)(C(=O)O)N

Specifications

52-52-8
100.0
White to Beige
97%
C6H11NO2
MFCD00001381
14, I, 526
cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
NILQLFBWTXNUOE-UHFFFAOYSA-N
1-aminocyclopentane-1-carboxylic acid
2901
129.16
Crystalline Flakes or Powder
96.0
320°C to 322°C
Authentic
Glass bottle
H2NC5H8CO2H
5 g
15, 2727
Solubility in water: ca 5g/100mL
C1CCC(C1)(C(=O)O)N
129.16
CHEBI:40547
97%
1-Amino-1-cyclopentanecarboxylic acid, 97%
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-144-6

RTECSNumber : GY2625000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only