missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzothiophene-5-carbaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC24804CB
Specifications
1-Benzothiophene-5-carbaldehyde | |
95% | |
C9H6OS | |
250 mg | |
QHHRWAPVYHRAJA-UHFFFAOYSA-N | |
1-benzothiophene-5-carbaldehyde | |
139097 | |
95% |
10133-30-9 | |
Amber Glass Bottle | |
MFCD05663673 | |
benzo b thiophene-5-carbaldehyde, benzo b thiophene-5-carboxaldehyde, 5-formylbenzo b thiophene, 1-benzothiophene-5-carboxaldehyde, benzothiophene-5-carbaldehyde, 5-thionaphthenecarbaldehyde | |
O=CC1=CC=C2SC=CC2=C1 | |
162.21 | |
162.21 |
Chemical Identifiers
10133-30-9 | |
162.21 | |
QHHRWAPVYHRAJA-UHFFFAOYSA-N | |
139097 | |
O=CC1=CC=C2SC=CC2=C1 |
C9H6OS | |
MFCD05663673 | |
benzo b thiophene-5-carbaldehyde, benzo b thiophene-5-carboxaldehyde, 5-formylbenzo b thiophene, 1-benzothiophene-5-carboxaldehyde, benzothiophene-5-carbaldehyde, 5-thionaphthenecarbaldehyde | |
1-benzothiophene-5-carbaldehyde |
Safety and Handling
Warning