missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzothiophene-5-carbaldehyde, 97%, Thermo Scientific™

$179.33 - $445.23

Chemical Identifiers

CAS 10133-30-9
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD05663673
InChI Key QHHRWAPVYHRAJA-UHFFFAOYSA-N
Synonym benzo b thiophene-5-carbaldehyde, benzo b thiophene-5-carboxaldehyde, 5-formylbenzo b thiophene, 1-benzothiophene-5-carboxaldehyde, benzothiophene-5-carbaldehyde, 5-thionaphthenecarbaldehyde
PubChem CID 139097
IUPAC Name 1-benzothiophene-5-carbaldehyde
SMILES O=CC1=CC=C2SC=CC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC24804CB
SDS View Documents
Thermo Scientific Chemicals
CC24804CB
250 mg Amber Glass Bottle
Each for $179.33
 
CC24804DA
SDS View Documents
Thermo Scientific Chemicals
CC24804DA
1 g Amber Glass Bottle
Each for $445.23
 
Description

Description

Specifications

Chemical Identifiers

10133-30-9
162.21
QHHRWAPVYHRAJA-UHFFFAOYSA-N
139097
O=CC1=CC=C2SC=CC2=C1
C9H6OS
MFCD05663673
benzo b thiophene-5-carbaldehyde, benzo b thiophene-5-carboxaldehyde, 5-formylbenzo b thiophene, 1-benzothiophene-5-carboxaldehyde, benzothiophene-5-carbaldehyde, 5-thionaphthenecarbaldehyde
1-benzothiophene-5-carbaldehyde

Specifications

10133-30-9
Amber Glass Bottle
MFCD05663673
benzo b thiophene-5-carbaldehyde, benzo b thiophene-5-carboxaldehyde, 5-formylbenzo b thiophene, 1-benzothiophene-5-carboxaldehyde, benzothiophene-5-carbaldehyde, 5-thionaphthenecarbaldehyde
O=CC1=CC=C2SC=CC2=C1
162.21
162.21
1-Benzothiophene-5-carbaldehyde
95%
C9H6OS
250 mg
QHHRWAPVYHRAJA-UHFFFAOYSA-N
1-benzothiophene-5-carbaldehyde
139097
95%
Videos
Safety and Handling

Safety and Handling

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'