missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzothiophene-5-carbonitrile, ≥95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC24816DA

 View more versions of this product

Catalog No. CC24816DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-Benzothiophene-5-carbonitrile
94.5
95%
C9H5NS
1g
IPJUOWGTGHKFKN-UHFFFAOYSA-N
1-benzothiophene-5-carbonitrile
2795452
95%
2060-63-1
100.0
Amber glass bottle
MFCD05664404
benzo b thiophene-5-carbonitrile, 5-cyanobenzothiophene, pubchem9722, 5-thionaphthenecarbonitrile, benzo thiophene-5-carbonitrile, benzo, a thiophene-5-carbonitrile, benzothiophene-5-carbonitrile, 1-benzo b thiophene-5-carbonitrile
N#CC1=CC=C2SC=CC2=C1
159.21
159.21

Chemical Identifiers

2060-63-1
159.21
IPJUOWGTGHKFKN-UHFFFAOYSA-N
2795452
N#CC1=CC=C2SC=CC2=C1
C9H5NS
MFCD05664404
benzo b thiophene-5-carbonitrile, 5-cyanobenzothiophene, pubchem9722, 5-thionaphthenecarbonitrile, benzo thiophene-5-carbonitrile, benzo, a thiophene-5-carbonitrile, benzothiophene-5-carbonitrile, 1-benzo b thiophene-5-carbonitrile
1-benzothiophene-5-carbonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions