missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzothiophene-5-carbonitrile, ≥95%, Thermo Scientific™

Chemical Identifiers

CAS 2060-63-1
Molecular Formula C9H5NS
Molecular Weight (g/mol) 159.21
MDL Number MFCD05664404
InChI Key IPJUOWGTGHKFKN-UHFFFAOYSA-N
Synonym benzo b thiophene-5-carbonitrile, 5-cyanobenzothiophene, pubchem9722, 5-thionaphthenecarbonitrile, benzo thiophene-5-carbonitrile, benzo, a thiophene-5-carbonitrile, benzothiophene-5-carbonitrile, 1-benzo b thiophene-5-carbonitrile
PubChem CID 2795452
IUPAC Name 1-benzothiophene-5-carbonitrile
SMILES N#CC1=CC=C2SC=CC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC24816CB
SDS View Documents
Thermo Scientific Chemicals
CC24816CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC24816DA
SDS View Documents
Thermo Scientific Chemicals
CC24816DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2060-63-1
159.21
IPJUOWGTGHKFKN-UHFFFAOYSA-N
2795452
N#CC1=CC=C2SC=CC2=C1
C9H5NS
MFCD05664404
benzo b thiophene-5-carbonitrile, 5-cyanobenzothiophene, pubchem9722, 5-thionaphthenecarbonitrile, benzo thiophene-5-carbonitrile, benzo, a thiophene-5-carbonitrile, benzothiophene-5-carbonitrile, 1-benzo b thiophene-5-carbonitrile
1-benzothiophene-5-carbonitrile

Specifications

2060-63-1
100.0
Amber glass bottle
MFCD05664404
benzo b thiophene-5-carbonitrile, 5-cyanobenzothiophene, pubchem9722, 5-thionaphthenecarbonitrile, benzo thiophene-5-carbonitrile, benzo, a thiophene-5-carbonitrile, benzothiophene-5-carbonitrile, 1-benzo b thiophene-5-carbonitrile
N#CC1=CC=C2SC=CC2=C1
159.21
159.21
1-Benzothiophene-5-carbonitrile
94.5
95%
C9H5NS
250mg
IPJUOWGTGHKFKN-UHFFFAOYSA-N
1-benzothiophene-5-carbonitrile
2795452
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'