missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
1-Benzothiophene-7-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS |
10134-98-2 |
Molecular Formula |
C9H6O2S |
Molecular Weight (g/mol) |
178.205 |
InChI Key |
LJPSRTWIAXVPIS-UHFFFAOYSA-N |
Synonym |
benzo b thiophene-7-carboxylic acid, benzo b thiophene-7-carboxylicacid, 7-carboxybenzo b thiophene, 1-benzo b thiophene-7-carboxylic acid |
PubChem CID |
14235073 |
IUPAC Name |
1-benzothiophene-7-carboxylic acid |
SMILES |
C1=CC2=C(C(=C1)C(=O)O)SC=C2 |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
10134-98-2 |
|
178.205 |
|
benzo b thiophene-7-carboxylic acid, benzo b thiophene-7-carboxylicacid, 7-carboxybenzo b thiophene, 1-benzo b thiophene-7-carboxylic acid |
|
1-benzothiophene-7-carboxylic acid |
|
C9H6O2S |
|
LJPSRTWIAXVPIS-UHFFFAOYSA-N |
|
14235073 |
|
C1=CC2=C(C(=C1)C(=O)O)SC=C2 |
Specifications
|
10134-98-2 |
|
Amber Glass Bottle |
|
1 g |
|
LJPSRTWIAXVPIS-UHFFFAOYSA-N |
|
1-benzothiophene-7-carboxylic acid |
|
14235073 |
|
97% |
|
97% |
|
C9H6O2S |
|
benzo b thiophene-7-carboxylic acid, benzo b thiophene-7-carboxylicacid, 7-carboxybenzo b thiophene, 1-benzo b thiophene-7-carboxylic acid |
|
C1=CC2=C(C(=C1)C(=O)O)SC=C2 |
|
178.205 |
|
178.21 |
|
1-Benzothiophene-7-carboxylic acid |
SDS
missing translation for 'documents'