missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl-3-pyrroline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 436770050

 View more versions of this product

Catalog No. AC436770050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Benzyl-3-pyrroline
96.0
0.8370g/mL
92°C
96% min. (GC)
5g
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
C1C=CCN1CC2=CC=CC=C2
159.23
159.23
6913-92-4
100.0
92°C (1.0 hPa)
Authentic
C11H13N
0.837
LRFHKHHUKGZIGE-UHFFFAOYSA-N
1-benzyl-2,5-dihydropyrrole
561506
97%

Chemical Identifiers

6913-92-4
159.23
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
1-benzyl-2,5-dihydropyrrole
C11H13N
LRFHKHHUKGZIGE-UHFFFAOYSA-N
561506
C1C=CCN1CC2=CC=CC=C2
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only