missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl-3-pyrroline, 97%, Thermo Scientific™

Chemical Identifiers

CAS 6913-92-4
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
InChI Key LRFHKHHUKGZIGE-UHFFFAOYSA-N
Synonym 1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
PubChem CID 561506
IUPAC Name 1-benzyl-2,5-dihydropyrrole
SMILES C1C=CCN1CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC436770010
SDS View Documents
Thermo Scientific Chemicals
436770010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436770050
View Documents
Thermo Scientific Chemicals
436770050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6913-92-4
159.23
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
1-benzyl-2,5-dihydropyrrole
C11H13N
LRFHKHHUKGZIGE-UHFFFAOYSA-N
561506
C1C=CCN1CC2=CC=CC=C2

Specifications

6913-92-4
100.0
92°C (1.0 hPa)
Authentic
Glass Bottle
1g
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
C1C=CCN1CC2=CC=CC=C2
159.23
159.23
1-Benzyl-3-pyrroline
96.0
0.8370g/mL
92°C
96% min. (GC)
C11H13N
0.837
LRFHKHHUKGZIGE-UHFFFAOYSA-N
1-benzyl-2,5-dihydropyrrole
561506
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only