Learn More
1-[(Benzyloxy)carbonyl]-3-piperidinecarboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC24701EB
Specifications
1-[(Benzyloxy)carbonyl]-3-piperidinecarboxylic acid | |
96.5 | |
97% | |
MFCD02931282 | |
1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester | |
C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O | |
263.293 | |
263.29 |
78190-11-1 | |
100.0 | |
C14H17NO4 | |
25g | |
FFLPIVZNYJKKDM-UHFFFAOYSA-N | |
1-phenylmethoxycarbonylpiperidine-3-carboxylic acid | |
234339 | |
97% |
Chemical Identifiers
78190-11-1 | |
263.293 | |
FFLPIVZNYJKKDM-UHFFFAOYSA-N | |
234339 | |
C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
C14H17NO4 | |
MFCD02931282 | |
1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester | |
1-phenylmethoxycarbonylpiperidine-3-carboxylic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.