missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
1-[(Benzyloxy)carbonyl]-3-piperidinecarboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS
78190-11-1
Molecular Formula
C14H17NO4
Molecular Weight (g/mol)
263.293
MDL Number
MFCD02931282
InChI Key
FFLPIVZNYJKKDM-UHFFFAOYSA-N
Synonym
1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester
PubChem CID
234339
IUPAC Name
1-phenylmethoxycarbonylpiperidine-3-carboxylic acid
SMILES
C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
CAS
78190-11-1
Molecular Weight (g/mol)
263.293
InChI Key
FFLPIVZNYJKKDM-UHFFFAOYSA-N
PubChem CID
234339
SMILES
C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
Molecular Formula
C14H17NO4
MDL Number
MFCD02931282
Synonym
1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester
IUPAC Name
1-phenylmethoxycarbonylpiperidine-3-carboxylic acid
Specifications
CAS
78190-11-1
CAS Max %
100.0
Packaging
Amber glass bottle
MDL Number
MFCD02931282
Synonym
1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester
SMILES
C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
Molecular Weight (g/mol)
263.293
Formula Weight
263.29
Chemical Name or Material
1-[(Benzyloxy)carbonyl]-3-piperidinecarboxylic acid
CAS Min %
96.5
Assay Percent Range
97%
Molecular Formula
C14H17NO4
Quantity
1g
InChI Key
FFLPIVZNYJKKDM-UHFFFAOYSA-N
IUPAC Name
1-phenylmethoxycarbonylpiperidine-3-carboxylic acid
PubChem CID
234339
Percent Purity
97%
Videos
Safety and Handling
Safety and Handling
GHS H Statement: Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'