missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Boc-amino)cyclopentanecarboxylic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1948303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
131°C to 134°C | |
35264-09-6 | |
MFCD01076126 | |
boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid | |
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O | |
229.276 | |
229.28 | |
1 g |
1-(Boc-amino)cyclopentanecarboxylic acid | |
C11H19NO4 | |
2734412 | |
YBZCSKVLXBOFSL-UHFFFAOYSA-N | |
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid | |
2734645 | |
98% |
Chemical Identifiers
35264-09-6 | |
229.276 | |
YBZCSKVLXBOFSL-UHFFFAOYSA-N | |
2734645 | |
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O |
C11H19NO4 | |
MFCD01076126 | |
boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid | |
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only